missing translation for 'onlineSavingsMsg'
Learn More

NE 10790, MedChemExpress

Product Code. 16493017
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493017 50 mg 50mg
16487036 100 mg 100mg
16479507 5 mg 5mg
16426968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493017 Supplier Medchem Express Supplier No. HY16011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent.

NE 10790, a poor farnesyl pyrophosphate synthase inhibitor, is a phosphonocarboxylate analogue of the potent bisphosphonate risedronate and is a weak antiresorptive agent.

Specifications

Chemical Name or Material NE 10790
Physical Form Solid
CAS 152831-36-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C8H10NO6P
Synonym 3-PEHPC
Solubility Information 0.1 M NaOH : 20 mg/mL (80.93 mM; ultrasonic and adjust pH to 6 with NaOH) ∣H2O : 5 mg/mL (20.23 mM; ultrasonic and adjust pH to 6 with NaOH)
SMILES OC(C(O)=O)(P(O)(O)=O)CC1=CC=CN=C1
Molecular Weight (g/mol) 247.14
Formula Weight 247.14
Percent Purity 98.28%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.