Learn More
Invitrogen™ NDUFV2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596678
Description
Immunogen sequence: MFFSAALRAR AAGLTAHWGR HVRNLHKTVM QNGAGGALFV HRDTPENNPD TPFDFTPENY KRIEAIVKNY PEGHKAAAVL PVLDLAQRQN GWLPISAMNK VAEVLQVPPM RVYEVATFYT MYNRKPVGKY HIQVCTTTPC MLRNSDSILE AIQKKLGIKV GETTPDKLFT LIEVECLGAC VNAPMVQIND NYYEDLTAKD IEEIIDELKA GKIPKPGPRS GRFSCEPAGG LTSLTEPPKG PGFGVQAGL; Positive Samples: MCF7, HT-29, PC-12, Mouse liver, Mouse brain, Rat liver, HL-60, NCI-H460, SKOV3, A-549, Mouse kidney, Mouse heart, Mouse testis, Rat kidney; Cellular Location: Mitochondrion inner membrane.
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFV2 | |
| Polyclonal | |
| Unconjugated | |
| NDUFV2 | |
| 24-kDa subunit of mitochondrial NADH dehydrogenase; 2900010C23Rik; CI-24k; complex I 24kDa subunit; complex I, mitochondrial respitory chain, 24 kD subunit; NADH dehydrogenase (ubiquinone) flavoprotein 2; NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa; NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial; NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit V2; NADH-ubiquinone oxidoreductase 24 kDa subunit; NADH-ubiquinone oxidoreductase flavoprotein 2; NDUFV2; nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4729, 72900, 81728 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19234, P19404, Q9D6J6 | |
| NDUFV2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human NDUFV2 (NP_0665522). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.