missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38428-100ul
This item is not returnable.
View return policy
Description
NDUFV2 Polyclonal antibody specifically detects NDUFV2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| NDUFV2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CI-24k, mitochondrial respitory chain, 24 kD subunit, NADH dehydrogenase (ubiquinone) flavoprotein 2 (24kD), NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa, NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial, NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial, NADH-ubiquinone oxidoreductase 24 kDa subunit, NADH-ubiquinone oxidoreductase flavoprotein 2, nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human NDUFV2 (NP_066552.2).,, Sequence:, MFFSAALRARAAGLTAHWGRHVRNLHKTVMQNGAGGALFVHRDTPENNPDTPFDFTPENYKRIEAIVKNYPEGHKAAAVLPVLDLAQRQNGWLPISAMNKVAEVLQVPPMRVYEVATFYTMYNRKPVGKYHIQVCTTTPCMLRNSDSILEAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGL | |
| 100 μL | |
| metabolism | |
| 4729 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction