missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11238-1-AP-20UL
This item is not returnable.
View return policy
Description
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.Specifications
| NDUFV1 | |
| Polyclonal | |
| Unconjugated | |
| NDUFV1 | |
| CI 51kD, Complex I 51kD, NDUFV1, UQOR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17995, 293655, 4723 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49821, Q91YT0 | |
| Ndufv1 | |
| NDUFV1 Fusion Protein Ag1764 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction