Learn More
NDUFV1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560950
Description
Immunogen sequence: HDWRLKGSLS RGDWYKTKEI LLKGPDWILG EIKTSGLRGR GGAGFPTGLK WSFMNKPSDG RPKYLVVNAD EGEPGTCKDR EILRHDPHKL LEGCLVGGRA MGARAAYIYI RGEFYNEASN LQVAIREAYE AGLIGKNACG SGYD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
Specifications
| NDUFV1 | |
| Polyclonal | |
| Unconjugated | |
| NDUFV1 | |
| CI-51K; CI51KD; CI-51kD; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; complex I-51kD; mitochondrial NADH dehydrogenase (ubiquinone) 51kDa subunit; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) flavoprotein 1; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase flavoprotein 1; NADH dehydrogenase flavoprotein 1; 24 kDa (FP); NADH dehydrogenase ubiquinone flavoprotein 1; NADH:ubiquinone oxidoreductase core subunit V1; NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1; UQOR1; wu:fc01f01; wu:fc12f12; zgc:86620 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49821 | |
| NDUFV1 | |
| Recombinant protein corresponding to Human NDUFV1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only