missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25172-1-AP-20UL
This item is not returnable.
View return policy
Description
Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.Specifications
| NDUFS8 | |
| Polyclonal | |
| Unconjugated | |
| NDUFS8 | |
| CI 23kD, Complex I 23kD, NDUFS8, TYKY, TYKY subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225887, 4728 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00217 | |
| NDUFS8 | |
| NDUFS8 Fusion Protein Ag18531 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction