Learn More
NDUFS8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25172-1-AP-150UL
 View more versions of this product
Product Code. 16873725
  • £349.00 / 150µL
Description
Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.
Specifications
Specifications
NDUFS8
Polyclonal
Unconjugated
NDUFS8
CI 23kD, Complex I 23kD, NDUFS8, TYKY, TYKY subunit
Rabbit
Antigen Affinity Chromatography
RUO
225887, 4728
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00217
NDUFS8
NDUFS8 Fusion Protein Ag18531
150 μL
Primary
Human, Mouse