missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS8 Antibody (S09-2B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19712-100ul
This item is not returnable.
View return policy
Description
NDUFS8 Monoclonal antibody specifically detects NDUFS8 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFS8 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4728 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-2B2 | |
| Unconjugated | |
| CI-23k, CI-23kD, CI23KD, complex I 23kDa subunit, Complex I-23kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 8 (23kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial, NADH-ubiquinone oxidoreductase 23 kDa subunit, TYKY, TYKY subunit | |
| A synthetic peptide of human NDUFS8 (Uniprot # O00217) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction