missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15728-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is a subunit of one of the complexes that forms the mitochondrial respiratory chain. This protein is one of over 40 subunits found in complex I, the nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. This complex functions in the transfer of electrons from NADH to the respiratory chain, and ubiquinone is believed to be the immediate electron acceptor for the enzyme. Mutations in this gene cause Leigh syndrome due to mitochondrial complex I deficiency, a severe neurological disorder that results in bilaterally symmetrical necrotic lesions in subcortical brain regions.Specifications
| NDUFS7 | |
| Polyclonal | |
| Unconjugated | |
| NDUFS7 | |
| CI 20KD, Complex I 20kD, MY017, NDUFS7, PSST, PSST subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362837, 374291, 75406 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75251 | |
| Ndufs7 | |
| NDUFS7 Fusion Protein Ag8471 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction