missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106367
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NDUFS7.
This gene encodes a protein that is a subunit of one of the complexes that forms the mitochondrial respiratory chain. This protein is one of over 40 subunits found in complex I, the nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. This complex functions in the transfer of electrons from NADH to the respiratory chain, and ubiquinone is believed to be the immediate electron acceptor for the enzyme. Mutations in this gene cause Leigh syndrome due to mitochondrial complex I deficiency, a severe neurological disorder that results in bilaterally symmetrical necrotic lesions in subcortical brain regions.
Specifications
| NDUFS7 | |
| Polyclonal | |
| Unconjugated | |
| Ndufs7 | |
| 1010001M04Rik; CI-20; CI-20KD; complex I 20kDa subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; complex I-20kD; FLJ45860; FLJ46880; MGC120002; MY017; NADH dehydrogenase (ubiquinone) Fe-S protein 7; NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S7; NADH:ubiquinone oxidoreductase PSST subunit; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 20 kDa subunit; NADH-ubiquinone oxidoreductase Fe-S protein 7; NDUFS7; PSST; PSST subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362837, 374291, 75406 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75251, Q9DC70 | |
| Ndufs7 | |
| A synthesized peptide derived from human NDUFS7(Accession O75251), corresponding to amino acid residues G38-C88. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction