missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFS6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564161
This item is not returnable.
View return policy
Description
Immunogen sequence: TGEKVTHTGQ VYDDKDYRRI RFVGRQK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.
This gene encodes a subunit of the NADH:ubiquinone oxidoreductase, which is the first enzyme complex in the electron transport chain of mitochondria. This complex functions in the transfer of electrons from NADH to the respiratory chain. The subunit encoded by this gene is one of seven subunits in the iron-sulfur protein fraction. Mutations in this gene cause mitochondrial complex I deficiency, a disease that causes a wide variety of clinical disorders, including neonatal disease and adult-onset neurodegenerative disorders.
Specifications
| NDUFS6 | |
| Polyclonal | |
| Unconjugated | |
| NDUFS6 | |
| BC059730; CI-13kA; CI13KDA; CI-13kD-A; complex I 13kDa subunit A; complex I, mitochondrial respiratory chain, 13-kD subunit; complex I-13kD-A; IP13; Ip13dis; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) Fe-S protein 6; NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial; NADH dehydrogenase Fe-S protein 6; NADH ubiquinone oxidoreductase; NADH:ubiquinone oxidoreductase NDUFS6 subunit; NADH:ubiquinone oxidoreductase subunit S6; NADH-ubiquinone oxidoreductase 13 kDa-A subunit; Ndub13; NDUFS6; RATIp13dis; zgc:103774 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75380 | |
| NDUFS6 | |
| Recombinant protein corresponding to Human NDUFS6. Recombinant protein control fragment (Product #RP-103552). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction