missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ndufs4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Ndufs4 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231606
|
Novus Biologicals
NBP3-38256-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227976
|
Novus Biologicals
NBP3-38256-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ndufs4 Polyclonal antibody specifically detects Ndufs4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| Ndufs4 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4724 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AQDQ, AQDQmitochondrial, CI-18 kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 4 (18kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Qreductase), NADH-coenzyme Q reductase, 18-KD, NADH-ubiquinone oxidoreductase 18 kDa subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human Ndufs4 (NP_002486.1).,, Sequence:, MAAVSMSVVLRQTLWRRRAVAVAALSVSRVPTRSLRTSTWRLAQDQTQDTQLITVDEKLDITTLTGVPEEHIKTRKVRIFVPARNNMQSGVNNTKKWKMEFDTRERWENPLMGWASTADPLSNMVLTFSTKEDAVSFAEKNGWSYDIEERKVPKPKSKSYGANFSWNKRTRVSTK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title