missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFS3 Recombinant Rabbit Monoclonal Antibody (23GB2145), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553235
This item is not returnable.
View return policy
Description
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
Specifications
| NDUFS3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NDUFS3 | |
| 0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4722 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2145 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75489 | |
| NDUFS3 | |
| A synthesized peptide derived from human NDUFS3 (200-264AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction