missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51940
This item is not returnable.
View return policy
Description
NDUFS3 Polyclonal specifically detects NDUFS3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| NDUFS3 | |
| Polyclonal | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_004542.1 | |
| NDUFS3 | |
| Peptide with sequence C-DTRPTVRPRNDVAHK corresponding to internal region according to NP_004542.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CI-30, CI-30kD, complex I 30kDa subunit, Complex I-30kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 3 (30kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial, NADH dehydrogenase-ubiquinone 30 kDa subunit, NADH-ubiquinone oxidoreductase 30 kDa subunit | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4722 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction