missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NDUFS3 Antibody (1D6), Novus Biologicals™

Product Code. 18344877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344877 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344877 Supplier Novus Biologicals™ Supplier No. H00004722M02100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFS3 Monoclonal antibody specifically detects NDUFS3 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00617
Gene Alias CI-30, CI-30kD, complex I 30kDa subunit, Complex I-30kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 3 (30kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial, NADH dehydrogenase-ubiquinone 30 kDa subunit, NADH-ubiquinone oxidoreductase 30 kDa subunit
Host Species Mouse
Immunogen NDUFS3 (AAH00617, 1 a.a. ∽ 264 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAAAVARLWWRGILGASALTRGTGRPSVLLLPVRRESAGADTRPTVRPRNDVAHKQLSAFGEYVAEILPKYVQQVQVSCFNELEVCIHPDGVIPVLTFLRDHTNAQFKSLVDLTAVDVPTRQNRFEIVYNLLSLRFNSRIRVKTYTDELTPIESAVSVFKAANWYEREIWDMFGVFFANHPDLRRILTDYGFEGHPFRKDFPLSGYVELRYDDEVKRVVAEPVELAQEFRKFDLNSPWEAFPVYRQPPESLKLEAGDKKPDAK
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4722
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.