Learn More
Invitrogen™ NDUFS2 Recombinant Rabbit Monoclonal Antibody (24GB495)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557108
 View more versions of this product
Product Code. 30274957
  • £126.00 / 20µL
Description
NDUFS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NDUFS2
Recombinant Monoclonal
Unconjugated
Ndufs2
AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312; unnamed protein product
Rabbit
Affinity chromatography
RUO
226646, 289218, 4720
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB495
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75306, Q641Y2, Q91WD5
Ndufs2
A synthesized peptide derived from human NDUFS2 (aa 300-434).
20 μL
Primary
Human, Mouse, Rat