missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ndufs1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12701-100ul
This item is not returnable.
View return policy
Description
Ndufs1 Polyclonal antibody specifically detects Ndufs1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Ndufs1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| CI-75k, CI-75Kd, complex I 75kDa subunit, complex I, mitochondrial respiratory chain, 75-kD subunit, Complex I-75kD, EC 1.6.5.3, EC 1.6.99.3, MGC26839, mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit, NADH dehydrogenase (ubiquinone) Fe-S protein 1 (75kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Qreductase), NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial, PRO1304 | |
| Produced in rabbits immunized with E. coli-derived Human Ndufs1 fragment | |
| 100 μg | |
| Vision | |
| 4719 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction