missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB9 Recombinant Rabbit Monoclonal Antibody (24GB90)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557499
This item is not returnable.
View return policy
Description
NDUFB9 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
LYRM3, also known as NADH dehydrogenase (ubiquinone) 1 beta subcomplex 9 (NDUFB9), is a ubiquitously expressed LYR-motif containing protein. It has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome, which is characterized by branchial and renal abnormalities and heredity deafness disorders. Other than its LYR-motif, LYRM3 appears to have no functional or structural relationship to either LYRM1 or LYRM2.
Specifications
| NDUFB9 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ndufb9 | |
| 1190008J14Rik; B22; CI-B22; complex I B22 subunit; complex I-B22; EGK_19260; LYR motif-containing protein 3; LYRM3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH:ubiquinone oxidoreductase subunit B9; NADH-ubiquinone oxidoreductase B22 subunit; NDUFB9; UQOR22 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 299954, 4715, 66218 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB90 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CQJ8, Q9Y6M9 | |
| Ndufb9 | |
| A synthesized peptide derived from human NDUFB9 (aa 50-200). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur