missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97197-50ul
This item is not returnable.
View return policy
Description
NDUFB9 Polyclonal antibody specifically detects NDUFB9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NDUFB9 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22) | |
| Produced in rabbits immunized with E. coli-derived Human NDUFB9 fragment. | |
| 50 μg | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 4715 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction