missing translation for 'onlineSavingsMsg'
Learn More

NDUFB9 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18408572 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408572 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408572 Supplier Novus Biologicals Supplier No. NBP177138G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFB9 Polyclonal antibody specifically detects NDUFB9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB9
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22)
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human LYRM3. The immunogen is located within the last 50 amino acids of LYRM3. Amino Acid Squence: ESWEREVKQLQEETPPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4715
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.