Learn More
Invitrogen™ NDUFB8 Recombinant Rabbit Monoclonal Antibody (6T8G1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537969
Description
Positive Samples: HeLa, HepG2, RAW264, 7, THP-1, C6, Mouse kidney, Mouse heart, Rat kidney, Rat heart Immunogen sequence: MAVARAGVLG VQWLQRASRN VMPLGARTAS HMTKDMFPGP YPRTPEERAA AAKKYNMRVE DYEPYPDDGM GYGDYPKLPD RSQHERDPWY SWDQPGLRLN WGEPMHWHLD MYNRNRVDTS PTPVSWHVMC MQLFGFLAFM IFMCWVGDVY.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [UniProt].
Specifications
| NDUFB8 | |
| Recombinant Monoclonal | |
| 0.40 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95169, Q9D6J5 | |
| NDUFB8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human NDUFB8 (O95169). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6T8G1 | |
| Unconjugated | |
| NDUFB8 | |
| 19 kDa; 2900010I05Rik; AI987932; ASHI; CI-ASHI; complex I ASHI subunit; complex I-ASHI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase 1 beta subcomplex 8; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase subunit B8; NADH-ubiquinone oxidoreductase ASHI subunit; NDUFB8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 293991, 4714, 67264 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.