Learn More
NDUFB8 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Product Code. 18402449 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
Product Code. unitSize
18402449 0.10mL
1 options
Product Code. 18402449 Supplier Novus Biologicals Supplier No. NBP305965MFV610
Rabbit Polyclonal Antibody
NDUFB8 Polyclonal antibody specifically detects NDUFB8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Applications Western Blot, Immunocytochemistry