missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58738-25ul
This item is not returnable.
View return policy
Description
NDUFB6 Polyclonal specifically detects NDUFB6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NDUFB6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| B17, CI, CI-B17, complex I, mitochondrial respiratory chain, B17 subunit, Complex I-B17, MGC13675, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6 (17kD, B17), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6, 17kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6, NADH-ubiquinone oxidoreductase B17 subunit, NADH-ubiquinone oxidoreductase beta subunit, 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4712 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| NDUFB6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KKSRIFPGDTILETGEVIPPMKEFPDQH | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur