missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545913
This item is not returnable.
View return policy
Description
Peptide sequence: ARTINVYPNF RPTPKNSLMG ALCGFGPLIF IYYIIKTERD RKEKLIQEGK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 86%.
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFB4 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB4 | |
| 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4710 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95168 | |
| NDUFB4 | |
| Synthetic peptide directed towards the C-terminal of human NDUFB4 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction