missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12358-1-AP-20UL
This item is not returnable.
View return policy
Description
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria.Specifications
| NDUFB3 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB3 | |
| B12, CI B12, Complex I B12, NDUFB3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301427, 4709, 66495 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43676, Q9CQZ6 | |
| NDUFB3 | |
| NDUFB3 Fusion Protein Ag3022 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction