missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103946
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NDUFB3.
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria.
Specifications
| NDUFB3 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB3 | |
| 2700033I16Rik; AI415450; B12; CI-B12; complex I-B12; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH:ubiquinone oxidoreductase subunit B3; NADH-ubiquinone oxidoreductase B12 subunit; Ndufb3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 301427, 4709, 66495 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43676, Q9CQZ6 | |
| NDUFB3 | |
| A synthesized peptide derived from human NDUFB3(Accession O43676), corresponding to amino acid residues M1-A38. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction