missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562394
This item is not returnable.
View return policy
Description
Immunogen sequence: QVFQSEFFSG LMWFWILWRF WHDSEEVLGH FPYPDPSQWT DEELGIPPDD ED Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 79%.
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
Specifications
| NDUFB2 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB2 | |
| 1810011O01Rik; 8kDa; AGGG; AI325567; CI-AGGG; complex I AGGG subunit; complex I-AGGG; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2, mitochondrial; NADH:ubiquinone oxidoreductase subunit B2; NADH-ubiquinone oxidoreductase AGGG subunit; NDUFB2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4708 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95178 | |
| NDUFB2 | |
| Recombinant protein corresponding to Human NDUFB2. Recombinant protein control fragment (Product #RP-100093). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction