missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93342-0.02ml
This item is not returnable.
View return policy
Description
NDUFB11 Polyclonal antibody specifically detects NDUFB11 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFB11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CI-ESSS, complex I NP17.3 subunit, Complex I-ESSS, ESSS, FLJ20494, MGC111182, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial, NADH-ubiquinone oxidoreductase ESSS subunit, Neuronal protein 17.3, Np15, Np17.3, P17.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human NDUFB11 (NP_061929.2). MAAGLFGLSARRLLAAAATRGLPAARVRWESSFSRTVVAPSAVAGKRPPEPTTPWQEDPEPEDENLYEKNPDSHGYDKDPVLDVWNMRLVFFFGVSIILVLGSTFVAYLPDYRCTGCPRAWDGMKEWSRREAERLVKYREANGLPIMESNCFDPSKIQLPEDE | |
| 0.02 mL | |
| Stem Cell Markers | |
| 54539 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction