missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597999
This item is not returnable.
View return policy
Description
NDUFB10 Polyclonal Antibody for Western Blot, IHC (P), ELISA
NDUFB10 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. Complex I is composed of 45 different subunits.
Specifications
| NDUFB10 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB10 | |
| 0610011B04Rik; 22kDa; CI PDSW; CI-PDSW; Complex I PDSW; complex I PDSW subunit; complex I-PDSW; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10; NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3); NADH:ubiquinone oxidoreductase subunit B10; NADH-ubiquinone oxidoreductase PDSW subunit; NDUFB 10; NDUFB10; PDSW | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4716, 68342 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O96000, Q9DCS9 | |
| NDUFB10 | |
| Recombinant Human NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10 protein (1-172AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction