missing translation for 'onlineSavingsMsg'
Learn More

NDUFB10 Antibody (OTI1H6), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18431764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431764 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431764 Supplier Novus Biologicals Supplier No. NBP272915AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFB10 Monoclonal antibody specifically detects NDUFB10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB10
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1H6
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CI-PDSW, complex I PDSW subunit, Complex I-PDSW, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 (22kD, PDSW), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10, NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3), NADH-ubiquinone oxidoreductase PDSW subunit, PDSW
Host Species Mouse
Immunogen Full length recombinant protein of human NDUFB10 (NP_004539) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4716
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.