missing translation for 'onlineSavingsMsg'
Learn More

NDUFB1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18348533
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348533 20 μg 20µL
18310984 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348533 Supplier Bio-Techne Supplier No. NBP31550020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFB1 Polyclonal antibody specifically detects NDUFB1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CI-MNLLCI-SGDH, complex I MNLL subunit, Complex I-MNLL, MNLL, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1 (7kD, MNLL), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1, 7kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1, NADH-ubiquinone oxidoreductase MNLL subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human NDUFB1 (NP_004536.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4707
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.