missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFAF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583895
This item is not returnable.
View return policy
Description
Immunogen sequence: MGWSQDLFRA LWRSLSREVK EHVGTDQFGN KYYYIPQYKN WRGQTIREKR IVEAANKKEV DYEAGDIPTE WEAWIRRTRK TPP Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 75%.
Mimitin, a small mitochondrial protein, whose transcription is directly stimulated by c-Myc, is highly expressed in 80% of esophageal squamous cell carcinomas (ESCC). Suppression of Mimitin expression by RNA interference had no effect in cancerous cell lines such as human cervical carcinoma or hepatocarcinoma cell lines, but caused a decrease in cell proliferation in human glioblastoma, embryonic lung fibroblastic cells, and ESCC, suggesting Mimitin may play a special role in these types of cells. Mimitin expression is also regulated by MAPK kinases and IL-1, but not through the NF-kappa-B-related pathway. It will interact with the microtubular protein MAP1S and can affect the activities of caspase-3 and -7 in cells stimulated to develop apoptosis. Other experiments suggest that Mimitin also acts as a molecular chaperone for the assembly of the mitochondrial complex I.
Specifications
| NDUFAF2 | |
| Polyclonal | |
| Unconjugated | |
| Ndufaf2 | |
| 1810058I14Rik; B17.2L; B17.2-like; B172L; B172-like; C86051; mimitin; mimitin, mitochondrial; MMTN; Myc-induced mitochondria protein; Myc-induced mitochondrial protein; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2; NADH dehydrogenase (ubiquinone) complex I, assembly factor 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; NADH dehydrogenase 1 alpha subcomplex assembly factor 2; NADH:ubiquinone oxidoreductase complex assembly factor 2; NDUFA12L; NDUFA12-like protein; Ndufaf2; RGD1560158 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N183 | |
| Ndufaf2 | |
| Recombinant protein corresponding to Human NDUFAF2. Recombinant protein control fragment (Product #RP-104844). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction