missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFAF1 Recombinant Rabbit Monoclonal Antibody (JE47-18)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534849
This item is not returnable.
View return policy
Description
Positive Control: K562 cell, 293T, SKOV-3, human liver cancer tissue, human prostate cancer tissue, human kidney tissue, THP-1.
The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications
| NDUFAF1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y375 | |
| NDUFAF1 | |
| Recombinant protein within Human NDUFAF1 aa 1-190. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE47-18 | |
| Unconjugated | |
| NDUFAF1 | |
| 2410001M24Rik; CGI65; CGI-65; CIA30; complex I intermediate-associated protein 30, mitochondrial; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH:ubiquinone oxidoreductase complex assembly factor 1; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; Ndufaf1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 51103 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction