missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications
Specifications
| Antigen | NDUFAF1 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NDUFAF1 |
| Gene Accession No. | Q9CWX2, Q9Y375 |
| Gene Alias | CGI 65, CGI65, CIA30, NDUFAF1 |
| Gene Symbols | NDUFAF1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?