missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15181-1-AP-150UL
This item is not returnable.
View return policy
Description
The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.Specifications
| NDUFAF1 | |
| Polyclonal | |
| Unconjugated | |
| NDUFAF1 | |
| CGI 65, CGI65, CIA30, NDUFAF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296086, 51103, 69702 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CWX2, Q9Y375 | |
| NDUFAF1 | |
| NDUFAF1 Fusion Protein Ag7162 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction