missing translation for 'onlineSavingsMsg'
Learn More

NDUFAF1 Rabbit anti-Human, Clone: 4K8Y8, Novus Biologicals™

Código de producto. 18354624 Tienda Bio Techne Productos
Change view
Click to view available options
Quantity:
100 μg
20 μg
Tamaño de la unidad:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
18354624 100 μg 100µL
18364523 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18354624 Proveedor Novus Biologicals N.º de proveedor NBP316313100UL

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

NDUFAF1 Monoclonal antibody specifically detects NDUFAF1 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen NDUFAF1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4K8Y8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CGI-65, CIA30CGI65, complex I intermediate-associated protein 30, mitochondrial, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human NDUFAF1 (Q9Y375).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51103
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.