missing translation for 'onlineSavingsMsg'
Learn More

NDUFAF1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA558997

Additional Details : Weight : 0.02000kg

Product Code. 15706871

  • £427.00 / 100µL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: SYDWSQFNTL YLRVRGDGRP WMVNIKEDTD FFQRTNQMYS YFMFTRGGPY WQEVKIPFSK FFFSNRGRIR DVQHELPLDK I Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications

Specifications

NDUFAF1
Polyclonal
Unconjugated
NDUFAF1
2410001M24Rik; CGI65; CGI-65; CIA30; complex I intermediate-associated protein 30, mitochondrial; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH:ubiquinone oxidoreductase complex assembly factor 1; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; Ndufaf1
Rabbit
Antigen affinity chromatography
RUO
51103
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y375
NDUFAF1
Recombinant protein corresponding to Human NDUFAF1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only