missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFAF1 Monoclonal Antibody (K70037_11A8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555891
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: SQRKTEGDLQ GDHQKEVALD ITSSEEKPDV SFDKAIRDEA IYHFRLLKDE IVDHWRGPEG HPLHEVLLEQ AKVVWQFRGK EDLDKWTVTS DKTIGGRSEV FLKMGKNNQS ALLYGTLSSE
The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
Specifications
| NDUFAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9Y375 | |
| NDUFAF1 | |
| Recombinant human NDUFAF1 (immunogen range is 51-170). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K70037_11A8 | |
| Unconjugated | |
| NDUFAF1 | |
| 2410001M24Rik; CGI65; CGI-65; CIA30; complex I intermediate-associated protein 30, mitochondrial; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH:ubiquinone oxidoreductase complex assembly factor 1; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; Ndufaf1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 51103 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction