missing translation for 'onlineSavingsMsg'
Learn More

NDUFAB1 Antibody (2D10), Novus Biologicals™

Product Code. 18342488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342488 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342488 Supplier Novus Biologicals Supplier No. H00004706M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFAB1 Monoclonal antibody specifically detects NDUFAB1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen NDUFAB1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2D10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH58920
Gene Alias ACPMGC65095, acyl carrier protein, mitochondrial, CI-SDAP, complex I SDAP subunit, FASN2A, mitochondrial acyl carrier protein, NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1 (8kD, SDAP), NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa, NADH:ubiquinone oxidoreductase SDAP subunit, NADH-ubiquinone oxidoreductase 9.6 kDa subunit, SDAP
Host Species Mouse
Immunogen NDUFAB1 (AAH58920, 1 a.a. ~ 156 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASRVLSAYVSRLPAAFAPLPRVRMLAVARPLSTALCSAGTQTRLGTLQPALVLAQVPGRVTQLCRQYSDMPPLTLEGIQDRVLYVLKLYDKIDPEKLSVNSHFMKDLGLDSLDQVEIIMAMEDEFGFEIPYIDAEKLMCPQEIVDYIADKKDVYE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4706
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.