missing translation for 'onlineSavingsMsg'
Learn More

NDUFA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 29621-1-AP-20UL

 View more versions of this product

Product Code. 17629676

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NDUFA9 Polyclonal antibody apecifically detetects NDUFA9 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

NDUFA9
Polyclonal
Unconjugated
BC009311
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Western Blot, Immunohistochemistry, ELISA
550 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
NDUFA9
NDUFA9 fusion protein Ag31222
20 μL
4704
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.