missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583598
This item is not returnable.
View return policy
Description
Immunogen sequence: RVLSMSRSAI TAIATSVCHG PPCRQLHHAL MPHGKGGRSS VSGIVATVFG ATGFLGRYVV NHLGRMGSQV IIPYRCDKYD IMHLRPMGDL GQLLFLEWDA RDKDSIRRVV QHSNVVINLI GRDWETKNFD FEDVFVKIPQ AI Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 85%.
NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.
Specifications
| NDUFA9 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA9 | |
| 1010001N11Rik; CC6; CI39k; CI-39k; CI-39kD; complex I 39kDa subunit; complex I-39kD; EMBL:AAH91192.1, ECO:0000312; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH:ubiquinone oxidoreductase subunit A9; NADH-ubiquinone oxidoreductase 39 kDa subunit; NDUFA9; ndufa9 {ECO:0000312; NDUFS2L; RGD:1307307}; SDR22E1; short chain dehydrogenase/reductase family 22E, member 1; sperm flagella protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4704 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16795 | |
| NDUFA9 | |
| Recombinant protein corresponding to Human NDUFA9. Recombinant protein control fragment (Product #RP-102237). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction