missing translation for 'onlineSavingsMsg'
Learn More

NDUFA9 Monoclonal Antibody (20C11B11B11), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 459100

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 10553134

  • £469.00 / Each
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Purification is near homogeneity as judged by SDS-PAGE. The antibody was produced in vitro using hybridomas grown in serum-free medium, and then purified by biochemical fractionation. 459100 detects a band at approximately 36 kDa. Use human heart mitochondria as a positive control.

NDUFA9 Monoclonal antibody specifically detects NDUFA9 in Bovine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

NDUFA9
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide
P34943, Q16795, Q9DC69, Q5BK63
NDUFA9
Bovine Heart Complex I
100 μg
Primary
Bovine, Human, Mouse, Rat
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
20C11B11B11
Unconjugated
NDUFA9
1010001N11Rik; CC6; CI39k; CI-39k; CI-39kD; complex I 39kDa subunit; complex I-39kD; EMBL:AAH91192.1, ECO:0000312; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH:ubiquinone oxidoreductase subunit A9; NADH-ubiquinone oxidoreductase 39 kDa subunit; NDUFA9; ndufa9 {ECO:0000312; NDUFS2L; RGD:1307307}; SDR22E1; short chain dehydrogenase/reductase family 22E, member 1; sperm flagella protein 3
Mouse
IgG fraction
RUO
362440, 404188, 4704, 66108
4° C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers