missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PGIVELPTLE ELKVDEVKIS SAVLKAAAHH YGAQCDKPNK EFMLCRWEEK DPRRCLEEGK LVNKCALDFF RQIKRHCA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
Specifications
| Antigen | NDUFA8 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NDUFA8 |
| Gene Accession No. | P51970 |
| Gene Alias | 0610033L03Rik; Aa2-258; AW261656; CI-19KD; CI-PGIV; Complex I-19kD; complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH:ubiquinone oxidoreductase PGIV subunit; NADH:ubiquinone oxidoreductase subunit A8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NDUFA8; PGIV |
| Gene Symbols | NDUFA8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
