missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539269
This item is not returnable.
View return policy
Description
NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFA8 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA8 | |
| NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8, Complex I-19kD, CI-19kD, Complex I-PGIV, CI-PGIV, NADH-ubiquinone oxidoreductase 19 kDa subunit, NDUFA8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4702, 68375 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51970, Q9DCJ5 | |
| NDUFA8 | |
| A synthetic peptide derived from the internal region of human NDUFA8 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction