missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93802-0.02ml
This item is not returnable.
View return policy
Description
NDUFA8 Polyclonal antibody specifically detects NDUFA8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| NDUFA8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50-1:100 | |
| CI-19KD, CI-PGIV, complex I PGIV subunit, Complex I-19kD, Complex I-PGIV, MGC793, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8 (19kD, PGIV), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8, NADH:ubiquinone oxidoreductase PGIV subunit, NADH-ubiquinone oxidoreductase 19 kDa subunit, PGIV | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 33-172 of human NDUFA8 (NP_055037.1). GAQCDKPNKEFMLCRWEEKDPRRCLEEGKLVNKCALDFFRQIKRHCAEPFTEYWTCIDYTGQQLFRHCRKQQAKFDECVLDKLGWVRPDLGELSKVTKVKTDRPLPENPYHSRPRPDPSPEIEGDLQPATHGSRFYFWTK | |
| 0.02 mL | |
| Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4702 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction