missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26161
This item is not returnable.
View return policy
Description
NDUFA7 Polyclonal specifically detects NDUFA7 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| NDUFA7 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NDUFA7 | |
| Peptide with sequence C-PKLPVGPSHKLSNN corresponding to internal region according to NP_004992.2. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.03 - 0.1 ug/ml, Immunohistochemistry 4 - 8 ug/ml, Immunohistochemistry-Paraffin 4 - 8 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| B14.5a, CI-B14.5a, complex I B14.5a subunit, Complex I-B14.5a, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (14.5kD, B14.5a), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7, NADH-ubiquinone oxidoreductase subunit B14.5a | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4701 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction