missing translation for 'onlineSavingsMsg'
Learn More

NDUFA4L2 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech

Product Code. p-7227507 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17761269 20 μL 20µL
17751269 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17761269 Supplier Proteintech Supplier No. CL4881648020UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFA4L2 Polyclonal antibody specifically detects NDUFA4L2 in Human, Mouse, Rat samples. It is validated for FC (Intra)

Specifications

Antigen NDUFA4L2
Applications Flow Cytometry
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite 488
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC003377
Host Species Rabbit
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 56901
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.