missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA4L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106688
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NDUFA4L2.
NDUFA4L2 is a protein coding gene.
Specifications
| NDUFA4L2 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA4L2 | |
| BC064011; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NDUFA4, mitochondrial complex associated like 2; NDUFA4L2; NUOMS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100362331, 407790, 56901 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4FZG9, Q9NRX3 | |
| NDUFA4L2 | |
| A synthesized peptide derived from human NDUFA4L2(Accession Q9NRX3), corresponding to amino acid residues N54-F87. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction