missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA4L2 Antibody (0B11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43005
This item is not returnable.
View return policy
Description
NDUFA4L2 Monoclonal antibody specifically detects NDUFA4L2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| NDUFA4L2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0B11 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100 | |
| NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2, NADH:ubiquinone oxidoreductase MLRQ subunit homolog, NADH-ubiquinone oxidoreductase MLRQ subunit homolog, NUOMSFLJ26118 | |
| NADH dehydrogenase(ubiquinone) 1 alpha subcomplex, 4-like 2 - (Uniprot# Q9NRX3) | |
| 100 μg | |
| Cell Biology, Endocrinology, Neurodegeneration, Neuroscience | |
| 56901 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction