missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118589
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NDUFA4.
Cytochrome c oxidase (COX, complex IV) is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. Required for complex IV maintenance. {ECO:0000269; PubMed:22902835}.
Specifications
| NDUFA4 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA4 | |
| CI-9k; CI-MLRQ; Complex I 9kDa subunit; complex I-MLRQ; Cytochrome c oxidase subunit NDUFA4; EC 1.6.5.3; EC 1.6.99.3; MLRQ; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9 kDa, MLRQ); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4; NADH-ubiquinone oxidoreductase MLRQ subunit; NDUFA4; NDUFA4, mitochondrial complex associated | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17992, 4697, 681024 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00483, Q62425 | |
| NDUFA4 | |
| A synthesized peptide derived from human NDUFA4(Accession O00483), corresponding to amino acid residues N48-F81. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction